Learn More
TUTTNAUER Tiva 15 Washer Disinfector with Automatic Sliding Door
Product Code. 30058118 Shop All Tuttnauer Products
Change view
Click to view available options
Voltage:
380/400 V
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
30058118 380/400 V 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 30058118 Supplier TUTTNAUER Supplier No. TIVA151VL
  • ONLINE EXCLUSIVE PRICE
    39784.50€
    34871.00€ / 1 unités
    Économisez 4913.50€
    12% de réduction
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Provides a flexible turnkey solution for washing, disinfecting and drying diverse medical instruments.
  • Equipped with automatic sliding door/s and best suited for surgical and sterilization departments.
  • Can be loaded with up to 6 washing levels for a total of 18 DIN baskets.
  • Compatible with a large variety of washing trolleys and accessories.
Specifications
Display LCD
Material Stainless Steel
Product Type Glassware Washer with Drying System and Steam Condenser
Style Automatic Sliding Door
Voltage 380/400 V