Learn More
Tizanidine Hydrochloride, LoGiCal
Product Code. 30460321
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460321 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460321 Supplier LoGiCal Supplier No. LGCFOR0887.00
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tizanidine hydrochloride
CAS 64461-82-1
Analyte or Component Names Tizanidine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C9 H8 Cl N5 S . Cl H
Certifications/Compliance ISO 9001
Synonym 5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine monohydrochloride, 5-Chloro-4-(2-imidazolin-2-ylamino)-2,1,3-benzothiadiazole hydrochloride, AB 021, AN 021, DS 103-282, Sirdalud, Ternelin, Tizan, Tizanidine Hydrochloride, Tizpa, Zanaflex
InChI Formula InChI=1S/C9H8ClN5S.ClH/c10-5-1-2-6-8(15-16-14-6)7(5)13-9-11-3-4-12-9;/h1-2H,3-4H2,(H2,11,12,13);1H
Solution Type Neat
SMILES Cl.Clc1ccc2nsnc2c1NC3=NCCN3
IUPAC Name 5-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 290.17
Quantity 10 mg
Formula Weight 288.9956 g/mol