Learn More
Tofogliflozin, TRC
Product Code. 30452100
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452100 5 mg 5mg
30453411 25 mg 25mg
30453853 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30452100 Supplier Toronto Research Chemicals Supplier No. TRCT5287505MG
 Restricted Item
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 903565-83-3
Molecular Formula C22 H26 O6
Molecular Weight (g/mol) 386.44
Synonym (1S,3'R,4'S,5'S,6'R)-6-[(4-Ethylphenyl)methyl]-3',4',5',6'-tetrahydro-6'-(hydroxymethyl)spiro[isobenzofuran-1(3H),2'-[2H]pyran]-3',4',5'-triol, CSG 452
IUPAC Name (3'R,4'S,5'S,6'R)-5-[(4-ethylphenyl)methyl]-6'-(hydroxymethyl)spiro[1H-2-benzofuran-3,2'-oxane]-3',4',5'-triol
SMILES CCc1ccc(Cc2ccc3COC4(O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c3c2)cc1
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C22H26O6/c1-2-13-3-5-14(6-4-13)9-15-7-8-16-12-27-22(17(16)10-15)21(26)20(25)19(24)18(11-23)28-22/h3-8,10,18-21,23-26H,2,9,11-12H2,1H3/t18-,19-,20+,21-,22?/m1/s1
Formula Weight 386.1729
Chemical Name or Material Tofogliflozin