Learn More
MP Biomedicals Top of CryoPrep Adapter
Product Code. 15752432 Shop All MP Biomedicals Products
Change view
Click to view available options
For Use With (Equipment):
FastPrep-24™
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15752432 FastPrep-24™ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15752432 Supplier MP Biomedicals Supplier No. 116002528T
  • ONLINE EXCLUSIVE PRICE
    543.00€
    516.60€ / 1 pièces
    Économisez 26.40€
    4% de réduction
Expédition estimée: 03-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
FastPrep-24 replacement part.
  • Adapter type: Cryogenic
  • Sample throughput: Medium
  • Tube compatibility: 2 mL
  • For FastPrep-24
Specifications
Sample Volume (Metric) 2 mL
Description Sample throughput: Medium
For Use With (Application) Holding plate to secure tubes and adapter
For Use With (Equipment) FastPrep-24™
Holds 24 x 2 mL Tubes
Product Type Replacement Top Component