Learn More
Topiramate, MedChemExpress
Product Code. 16492026
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492026 500 mg 500mg
16458116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492026 Supplier Medchem Express Supplier No. HYB0122
 Restricted Item
Expédition estimée: 20-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
Topiramate (McN 4853) is a broad-spectrum antiepileptic agent. Topiramate is a GluR5 receptor antagonist. Topiramate produces its antiepileptic effects through enhancement of GABAergic activity, inhibition of kainate/AMPA receptors, inhibition of voltage-sensitive sodium and calcium channels, increases in potassium conductance, and inhibition of carbonic anhydrase.
Specifications
Chemical Name or Material Topiramate
CAS 97240-79-4
Color White
Purity Grade Notes Research
Molecular Formula C12H21NO8S
Quantity 500 mg
Synonym McN 4853 RWJ 17021
Solubility Information DMSO : ≥ 100 mg/mL (294.67 mM) ∣H2O : 4 mg/mL (11.79 mM; Need ultrasonic)
SMILES NS(OC[C@]1(OC(C)(C)O2)[C@@H]2[C@H](OC(C)(C)O3)[C@H]3CO1)(=O)=O
Molecular Weight (g/mol) 339.36
Formula Weight 339.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation