Learn More
PureAire Monitoring Systems, Inc. Trace Oxygen Analyser, 0-1000ppm
Product Code. 16208831
Change view
Click to view available options
Measurement Range:
0 to 1000 ppm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
16208831 0 to 1000 ppm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 16208831 Supplier PureAire Monitoring Systems, Inc. Supplier No. 99141
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
For use in a vacuum environment, 3D printer, nitrogen generator, or semiconductor tool using nitrogen, argon, or cryogenics. The robust zirconium sensor is connected via the KF25 flange. Sensor is connected to the analyser using a 3m cable for easy connection.
  • Accurate ppm Zirconium Sensor
  • Two-Alarm setpoints
  • Built-in audible alarm 90 db
  • Digital display, and 4-20mA analog output
  • Wall mounting brackets
Why choose PureAire?
  • As an industry leader in oxygen analyser and gas detection, PureAire is dedicated to the safety of our clients, and the customers they serve, by manufacturing monitors that are long-lasting, innovative, and reliable.
What is an O2 Analyser?
  • The Air Check Trace O2 Analyser is designed for continuous monitoring low oxygen levels need from zero to 1,000 parts per million (ppm)
What gases can an O2 monitor be used for?
  • PureAire’s oxygen analysers can be used for nitrogen, and argon.
Certifications
CE Marked
Warranty
2 years
Specifications
Depth (English) 3 in.
Depth (Metric) 76 mm
Dimensions (L x W x H) 5.125 x 3.15 x 3 in.
Height (English) 3.15 in.
Height (Metric) 80 mm
Humidity Range 0-95% RH
Measurement Range 0 to 1000 ppm
Temperature Range (English) -40°F to +134°F
Temperature Range (Metric) -40 to +55°C
Voltage 24V
Warranty 2 Year after ETD
Weight (English) 1.1 lb.
Weight (Metric) 0.5 kg
Width (English) 5.125 in.
Width (Metric) 130 mm