Description
- Type 5/6 lightweight and breathable coverall, offering protection against hazardous dusts and limited liquid splashes
- Standard collar provides comfort and convenience when a hood is not needed
- Knitted cuffs with elastic waist and ankles for convenience and freedom of movement
- Two-way zipper with storm flap for convenience and to help provide additional protection
- Anti-static coating on both sides helps reduce static build-up
- Robust, low-linting, highly breathable 43 gsm SMMMS material
- Meets inward leakage requirements of EN 1073-2 for protection against nuclear particles
Specifications
Specifications
| Product Type | Protective Coveralls |
| Size | Medium |
| Gender | Unisex |
| Material | SMS Fabric |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Cuff Style | Knit |
| Closure Type | Front Zipper |
| Seam Type | Serged |
| Show More |