Learn More
trans-p-Coumaric Acid, TRC
Product Code. 17914845
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17914845 10 g 10g
17994285 25 g 25g
17971375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17914845 Supplier Toronto Research Chemicals Supplier No. TRCC75539010G
 Restricted Item
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 501-98-4
Molecular Formula C9 H8 O3
Molecular Weight (g/mol) 164.16
Synonym 2-Propenoic acid, 3-(4-hydroxyphenyl)-, (2E)- (9CI, ACI), (2E)-3-(4-Hydroxyphenyl)-2-propenoic acid (ACI), 2-Propenoic acid, 3-(4-hydroxyphenyl)-, (E)- (ZCI), Cinnamic acid, p-hydroxy-, (E)- (8CI), (2E)-3-(4-Hydroxyphenyl)prop-2-enoic acid, (E)-3-(4-Hydroxyphenyl)-2-propenoic acid, (E)-3-(4-Hydroxyphenyl)acrylic acid, (E)-3-(4-Hydroxyphenyl)acrylic acid, (E)-4-Hydroxycinnamic acid, (E)-p-Coumaric acid, (E)-p-Coumaric acid, (E)-p-Hydroxycinnamic acid, Naringeninic acid, p-Hydroxy-trans-cinnamic acid, p-trans-Coumaric acid, trans-3-(4-Hydroxyphenyl)-2-propenoic acid, trans-4-Coumaric acid, trans-4-Hydroxycinnamic acid, trans-p-Coumaric acid, trans-p-Coumarinic acid, trans-p-Hydroxycinnamic acid
IUPAC Name (E)-3-(4-hydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\c1ccc(O)cc1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C9H8O3/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-6,10H,(H,11,12)/b6-3+
Formula Weight 164.0473
Percent Purity >95%
Chemical Name or Material trans-p-Coumaric acid
Grade HPLC