Learn More
Zeiss™ Transmitted Light Equipment S
Description
Compatible Microscopes:
- Axio Zoom.V16
- SteREO Discovery.V12
- SteREO Discovery.V20
- SteREO Discovery.V8
- SteREO Lumar.V12
Specifications
Specifications
| Type | Transmitted Light Equipment |
| Description | Transmitted light equipment |
| For Use With (Application) | Transmitted Light |
| For Use With (Equipment) | Stand Base 450 |
| Includes | Light guide and 2 shiftable and tiltable reflectors for bright field/dark field and for variable oblique light |