Learn More
SI Analytics™ Transparent Thermostat CT 72/4
Product Code. 11994719 Shop All SI Analytics Products
Change view
Click to view available options
Capacity (Metric):
28 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11994719 28 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11994719 Supplier SI Analytics™ Supplier No. 285418568
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Transparent thermostat CT 72/4 is used for viscosity measurement using glass capillary viscometers.
  • Working temperature 5°C to 150°C, stainless steel container with thermal insulation, for 4 measuring positions
  • Suitable for manual measurement or AVS™/S measuring stands
  • Heater with Pt 100-PID control
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 28 L
Temperature Range 30°C to 150°C
Includes Two Manual Gauge Slides
Material Stainless Steel
Temperature Stability ±0.02°C
Dimensions (L x W x H) 25 x 60.5 x 53.5 cm
Weight (Metric) 23 kg
Voltage 230 V