Learn More
Corning™ Transport Medium
BIOPHARMA
Product Code. 16681563
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
16681563 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16681563 Supplier Corning™ Supplier No. 25500CM
  • ONLINE EXCLUSIVE PRICE
    740.00€
    677.03€ / 6 pièces
    Économisez 62.97€
    8% de réduction
 Restricted Item
Expédition estimée: 30-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Corning™ Transport Medium is an isotonic, neutral pH solution that contains protective proteins, antibiotics to prevent microbial growth and contamination
  • This isotonic, neutral pH solution contains protective proteins/antibiotic to prevent microbial growth, and anti-mycotic to prevent contamination by yeast and fungi
  • For research and further manufacturing purposes only
  • Not for direct use in humans or animals.
  • Formulated using the final ingredient concentrations outlined in the CDC's Preparation of Viral Transport Medium (SOP#: DSR-052-01)
  • Formulation contains Hanks' Balanced Salt Solution with Ca and Mg (without phenol red), 2% Heat Inactivated Fetal Bovine Serum, 100 μg/mL Gentamicin and 0.5 μg/mL Amphotericin B
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Formulated in accordance to the CDC's Preparation of Viral Transport Medium guidelines
For Use With (Application) Research, Manufacturing Applications
Quantity 6/Cs.
Storage Requirements 2°C to 8°C
Volume (Metric) 1 L