Learn More
Applied Biosystems™ Trifluoroacetic Acid (TFA) Neat Liquid
Product Code. 15820273
Change view
Click to view available options
Form:
Liquid
Unit Size:
450mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Form Unit Size
15820273 Liquid 450mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15820273 Supplier Applied Biosystems™ Supplier No. 400137
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Trifluoroacetic Acid (TFA, concentrated) for use with OPC™ cartridges for oligonucleotide purification.
  • For Research Use Only
  • Not for use in diagnostics procedures
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type Trifluoroacetic Acid (TFA) Neat Liquid
Chemical Name or Material trifluoroacetic acid
Shipping Condition Room Temperature
Reagent Type Solvent or General Reagent
Format Liquid