Learn More
Trifluoroiodomethane, 97+%
Product Code. 11374218
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374218 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an ozone depleters according to Regulation (EC) No 1005/2009, all customers must complete a declaration before delivery.
Product Code. 11374218 Supplier Thermo Scientific Alfa Aesar Supplier No. 041479.14
 Restricted Item
Expédition estimée: 14-12-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an ozone depleters according to Regulation (EC) No 1005/2009, all customers must complete a declaration before delivery.
CAS: 2314-97-8 | CF3I | 195.911 g/mol
Trifluoroiodomethane is used as a gaseous fire suppression flooding agent for in-flight aircraft and electronic equipment fires. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is involved in the rhodium-catalyzed alfa-trifluoromethylation of alfa,beta-unsaturatedketones. It plays an important role as catalyst in the enantioselective alfa -trifluoromethylation of aldehydes through photoredox organocatalysis using a readily available iridium photocatalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2314-97-8
Molecular Formula CF3I
Molecular Weight (g/mol) 195.911
MDL Number MFCD00001060
InChI Key VPAYJEUHKVESSD-UHFFFAOYSA-N
Synonym trifluoromethyl iodide, iodotrifluoromethane, methane, trifluoroiodo, perfluoromethyl iodide, trifluoro iodo methane, trifluoromethyliodide, monoiodotrifluoromethane, freon 13t1, unii-42a379kb0u, ccris 7782
PubChem CID 16843
IUPAC Name trifluoro(iodo)methane
SMILES C(F)(F)(F)I
Specifications
Density 2.361 g/mL
Boiling Point -22.5°C
Flash Point None
Packaging Cylinder
Refractive Index 1.379
Quantity 25 g
UN Number UN1956
Beilstein 1732740
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 195.91
Percent Purity ≥97%
Chemical Name or Material Trifluoroiodomethane