Learn More
Trolox, MedChemExpress
Product Code. 16480759
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480759 1 g 1g
16446097 5 g 5g
16401536 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16480759 Supplier Medchem Express Supplier No. HY101445
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MedChemExpress Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage.
Trolox is an analogue of vitamin E with a powerful antioxidant effect. Trolox is also a powerful inhibitor of membrane damage.
Specifications
Chemical Name or Material Trolox
CAS 53188-07-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H18O4
Quantity 1 g
Solubility Information DMSO : ≥ 100 mg/mL (399.54 mM)
SMILES O=C(C1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1)O
Molecular Weight (g/mol) 250.29
Formula Weight 250.29
Percent Purity 98.19%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death