Learn More
Edulab Tuning Forks, Aluminum, Set of 8
Product Code. 30280288
Change view
Click to view available options
Product Type:
Tuning Fork Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30280288 Tuning Fork Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280288 Supplier Edulab Supplier No. VTN12302576
  • ONLINE EXCLUSIVE PRICE
    29.70€
    28.73€ / 1 kits
    Économisez 0.97€
    3% de réduction
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Tuning fork scientifically tuned to the absolute concert pitch middle C.
  • Tuning fork scientifically tuned to the absolute concert pitch middle C.(256 Hz)
  • Aluminum construction ensures durability and long-lasting use.
  • Set of 8 forks included for multiple experiments or applications.
  • Two-pronged plain shank fork used for tuning musical instruments, calculating frequency in scientific experiments, understanding the principles of sound waves, and in chakra healing practices.
  • Used as an acoustic resonator. Resonates at the specific constant pitch after striking against a surface and emits a pure musical tone once the high overtones fade out.
Specifications
Product Type Tuning Fork Set