Learn More
Corning Universal power supply 9V with set of adaptors EU,
Brand:  Corning 4996
Universal power supply 9V with set of adaptors EU, UK, US, AUS
Product Code. 18888880
  • ONLINE EXCLUSIVE PRICE
    43.60€
    40.73€ / 1 pièces
    Économisez 2.87€
    6% de réduction
 Restricted Item
Expédition estimée: 02-04-2027