Learn More
Tyrosinase-Related Protein-1 Rabbit, Clone: TYRP1/1564R, enQuireBio™
Product Code. 15981038
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15981038 20 μg 20µg
15991038 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15981038 Supplier EnquireBio Supplier No. 7306RBM4P0
Please to purchase this item. Need a web account? Register with us today!
This product has been withdrawn. Where a replacement item has been identified it will be shown below
View alternative products
This item is not returnable. View return policy
Rabbit Monoclonal Antibody
Specifications
Antigen Tyrosinase-Related Protein-1
Applications Immunohistochemistry
Cellular Localization Cytoplasmic
Classification Monoclonal
Clone TYRP1/1564R
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with or without 0.05% BSA and 0.05% azide.
Gene Accession No. NP_000541
Gene Symbols TYRP1
Host Species Rabbit
Immunogen Recombinant human full-length TYRP1 protein
Molecular Weight of Antigen 75kDa
Positive Control SK-MEL-23, SK-MEL-19, SK-MEL-30, SK-MEL-37 cells or Melanoma.
Purification Method Protein A/G Purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7306
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG κ