Learn More
SI Analytics™ Ubbelohde Viscometers for Manual Measurements
Product Code. 12008163 Shop All SI Analytics Products
Change view
Click to view available options
Viscosity Range:
0.8 to 5 cSt
50 to 500 cSt
Approximate Constant:
0.005
0.5
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Viscosity Range Approximate Constant Unit Size
12008163 0.8 to 5 cSt 0.005 Each
12058163 50 to 500 cSt 0.5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12008163 Supplier SI Analytics™ Supplier No. 285400029
  • ONLINE EXCLUSIVE PRICE
    708.00€
    513.00€ / 1 pièces
    Économisez 195.00€
    27% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
SI Analytics™ Ubbelohde Viscometers for Manual Measurements feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.
  • Calibrated, with constant, for manual measurement
  • Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, and high-temperature or low temperature measurements
  • Delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
  • All viscometers are provided with ring marks
  • This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
  • The recommended minimum flowthrough time is 200 seconds
TRUSTED_SUSTAINABILITY
Specifications
Product Type Ubbelohde Viscometer
Viscosity Range 0.8 to 5 cSt
Calibrated Yes
Description 0.53 mm diameter
Approximate Constant 0.005
Certifications/Compliance DIN 51 562 Part 1, ISO/DIS 3105 (BS-IP-SL)
Material Glass
Length (Metric) 290 mm
For Use With (Application) Manual measurements
Volume (Metric) 15 to 20 mL