Learn More
Thermo Scientific™ UltiMate™ 3000 Electrochemical Detector Sensors
Shop All Thermo Scientific ProductsDescription
Choose either a coulometric or an amperometric analytical Thermo Scientific™ Dionex™ UltiMate™ 3000 Electrochemical Detector Sensor for your electrochemical detector. They offer both high sensitivity and selectivity for your target analytes.
Specifications
Specifications
| Detection Mode | DC/PAD |
| Detector Type | Electrochemical (ECD) |
| Type | 6041RS Ultra Amperometric Analytical Sensor with Accessories and Gaskets |
| For Use With (Application) | Amperometric Detection |
| For Use With (Equipment) | UltiMate 3000 Amperometric Detector |
| No. of Channels | 1 |
| Reference Electrode | Pd |
| Working Electrodes | GC, BDD, Au |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?