Learn More
CleanSpace Technology™ Ultra Decontamination Plug Set
Product Code. 15950072 Shop All CleanSpace Technology Products
Change view
Click to view available options
For Use With (Equipment):
CleanSpace™ Ultra Respirator unit (PAF-0070)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15950072 CleanSpace™ Ultra Respirator unit (PAF-0070) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Do not wear the respirator when the plug set is in place.
Product Code. 15950072 Supplier CleanSpace Technology™ Supplier No. PAF0074
  • ONLINE EXCLUSIVE PRICE
    73.49€
    57.00€ / 1 pièces
    Économisez 16.49€
    22% de réduction
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Do not wear the respirator when the plug set is in place.
For use with the CleanSpace™ Ultra Respirator unit (PAF-0070). Seals all the openings on the CleanSpace™ Ultra PAPR unit for storage during and after decontamination. The Plug Set is made of nylon and silicone and is safe for use in decontamination operations.
  • Used with the revolutionary CleanSpace Ultra PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Material Nylon, Silicone
For Use With (Equipment) CleanSpace™ Ultra Respirator unit (PAF-0070)
Dimensions (L x W x H) 800 x 180 x 110 mm
Weight (Metric) 100 g