Learn More
WTW™ Ultrapure Water Conductivity Cell
Product Code. 10603732
Change view
Click to view available options
Diameter (Metric):
12 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10603732 12 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10603732 Supplier WTW™ Supplier No. 301961
  • ONLINE EXCLUSIVE PRICE
    1150.00€
    741.00€ / 1 pièces
    Économisez 409.00€
    35% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Obtain pure water measuring results with this ultrapure water conductivity cell that includes built-in temperature sensor, 4.9 ft (1.5m) cable and a waterproof plug.
  • Measuring cells with concentric electrodes of stainless steel
  • Flow vessels made of glass or stainless steel for measurements in pure water
  • Ultrapure water conductivity cell
  • Includes built-in temperature sensor, 4.9 ft (1.5m) cable and waterproof plug
  • Precise measured values thanks to concentric electrodes without edge fields
  • Includes flow vessels to prevent wrong results due to by carbon dioxide
  • No polarization effects thanks to stainless steel design 
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) 0.47 in.
Description Ultrapure water conductivity cell
Diameter (Metric) 12 mm
Includes Built-in temperature sensor, 4.9 ft (1.5m) cable and waterproof plug