Learn More
Biochrom™ Ultrospec 7500 UV-Vis Spectrophotometer
Product Code. 17717359 Shop All Harvard Bioscience Products
Change view
Click to view available options
Type:
Local Control UV-Vis Spectrophotometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17717359 Local Control UV-Vis Spectrophotometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17717359 Supplier Biochrom™ Supplier No. 80214060
  • ONLINE EXCLUSIVE PRICE
    11285.00€
    9739.80€ / 1 pièces
    Économisez 1545.20€
    13% de réduction
View all promotions
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
The Ultrospec 7500 is a simple-to-use instrument with highly advanced performance functionality and a sophisticated GLP mode to ensure high quality, consistent and reliable results. Versatile and precise, this instrument is ideal for use in any manufacturing or production laboratory.
  • Easy to use icon driven touch screen for true 'plug and play' setup
  • Automated 8-cell sample carousel supplied as standard
  • Robust, compact and reliable for general or heavily regulated laboratory use
  • Combines basic measurement modes with enhances software functionality and method development capabilities
  • Can be upgraded with Resolution CFR PC control software to comply with 21 CFR Part 11
  • Provides stored methods for DNA, RNA, oligo and protein quantification, as well as checking sample purity
  • Preloaded methods for commonly available fluorescent dyes for previous samples
  • Screenshot function to easily capture and export results
  • 800 x 480 pixel backlit, color LCD touch display
  • Xenon flash lamp for longer lifetime and lower running costs
  • IQ/OQ available on request
Specifications
Absorbance Range 190 to 1100 nm
Baseline Flatness ±0.002 Peak to Peak, ±0.0005 RMS at 340 nm for 0 A
Beam Center Height 15 mm
Color White
Data Interval for Scanning 1 nm
Detector Type Photodiode
Display LCD
Dimensions (L x W x H) 510 × 350 × 160 mm
Languages English, French, German, Spanish, Italian, Chinese, Japanese
Light Source Pulsed Xenon
Product Line Ultrospec 7000
Spectral Bandwidth <2 nm
Type Local Control UV-Vis Spectrophotometer
Voltage 100/240 V
Wavelength Range 190 to 1100 nm
Wavelength Selection 1 nm
Electrical Requirements 100 to 240 V
Monochromator 1200 Lines/mm Aberration Corrected Concave Grating
Optical Design Split Beam Optics
Photometric Accuracy Instrument ±0.5% or ±0.003 A (Whichever Is Greater at 546 nm)
Photometric Range -3 to +3 A, 0.1 to 100% T
Photometric Repeatability ±0.5% to 3 A at 546 nm
Printer Included Printer Accessory Available
Shape Rectangular
Warranty 2 Year Instrument, 3 Year Lamp
Wavelength Accuracy ±1 nm
Wavelength Repeatability ±0.5 nm
Frequency 50/60 Hz