Learn More
Uridine 5’-Diphosphate Sodium Salt, TRC
Spécification
Spécification
| Chemical Name or Material | Uridine 5'-Diphosphate Sodium Salt |
| CAS | 21931-53-3 |
| Molecular Formula | C9 H12 N2 O12 P2 . 2 Na |
| Quantity | 250 mg |
| Synonym | Uridine 5'-Diphosphate Sodium Salt |
| InChI Formula | InChI=1S/C9H14N2O12P2.2Na/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(22-8)3-21-25(19,20)23-24(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,10,12,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1 |
| SMILES | [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(=O)([O-])OP(=O)(O)[O-])N2C=CC(=O)NC2=O |
| IUPAC Name | disodium;[[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate |
| Molecular Weight (g/mol) | 448.12 |
| Formula Weight | 447.9661 |