missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Glass ACD Solution Tube with Yellow Closure

Product Code. 15167014
Click to view available options
Capacity (Metric):
6 mL
8.5 mL
Additive:
ACD Solution A
ACD Solution B
Closure Type:
Hemogard
Traditional Cap
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15167014

Brand: BD 367756

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to obtain a whole blood or a plasma sample. BD Vacutainer™ Glass ACD Solution Tube with Yellow Closure contains the active anticoagulant Tri-sodium Citrate with Citric Acid and Dextrose. Two formulations of the ACD solution are available.

BD Vacutainer™ ACD tubes are available in glass only and are identifiable by a yellow cap (Hemogard™ or conventional). They contain the active anticoagulant Tri-sodium Citrate with Citric Acid and Dextrose in one of two formulations. The concentration of additives in each solution is as follows:
  • ACD Solution A (conventional closure)
    • Sodium citrate: 22.0g/L
    • Dextrose: 24.5g/L
    • Citric acid: 8.0g/L
    • Antimycotic (K sorbate) reagent: 0.15g/L
  • ACD Solution B (Hemogard™ closure)
    • Sodium citrate: 13.2g/L
    • Dextrose: 14.7g/L
    • Citric acid: 4.8g/L
    • Antimycotic (K sorbate) reagent: 0.2g/L
TRUSTED_SUSTAINABILITY

Specifications

Product Type ACD Solution B Tube
Additive ACD Solution B
Material Glass
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 6 mL
Closure Color Yellow
Tube Size 13 x 100 mm
For Use With (Application) Used to obtain a whole blood or plasma sample
Label Type Paper
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.