Learn More
Hamilton™ VALVE, HV, 3-2-Y, PSD/4
Description
- CTFE (chlorotrifluorethylene) construction
- High compression strength
- Low gas permeability
- Mechanically, chemically, thermally resistant
- Suitable for PSD/4 compact syringe pump
Specifications
Specifications
| For Use With (Equipment) | PSD/4 Compact Syringe Pump |
| Type | 3-2-Y Valve |