Learn More
Testo™ Vane Anemometer
Product Code. 11381314
Change view
Click to view available options
Velocity:
0.4 to 20 m/sec.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Velocity Unit Size
11381314 0.4 to 20 m/sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11381314 Supplier Testo™ Supplier No. 0560 4101
  • ONLINE EXCLUSIVE PRICE
    143.91€
    116.00€ / 1 pièces
    Économisez 27.91€
    19% de réduction
Expédition estimée: 20-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Specifications
Type Vane Anemometer
Accuracy ±0.5 m/sec.
Dimensions (L x W x H) 133 x 46 x 25 mm
For Use With (Application) Measures air flow and air temperature
Temperature (English) 14°F to 122°F
Temperature (Metric) -10°C to +50°C
Velocity 0.4 to 20 m/sec.