Learn More
Thermo Scientific™ Vanquish™ Charged Aerosol Detectors
Product Code. 17871266 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
Vanquish CAD F
Vanquish CAD H
Temperature Range:
35°C, 50°C, 70°C
Ambient +5° to +100°C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Temperature Range Unit Size
17871266 Vanquish CAD H Ambient +5° to +100°C Each
17575456 Vanquish CAD F 35°C, 50°C, 70°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17871266 Supplier Thermo Scientific™ Supplier No. VHD20A
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Performance you can see. Confidently detect and quantify all non-volatile and many semi-volatile analytes with Thermo Scientific™ Vanquish™ Charged Aerosol Detectors.
Expand your detection range and get accurate quantitation in your high-performance liquid chromatography (HPLC) analysis with the near-universal Charged Aerosol Detector (CAD). Confidently quantify non-volatile and many semi-volatile analytes. Thermo Scientific™ Vanquish™ CADs deliver sensitive and consistent response across compound classes, helping you accurately measure both low-level impurities and highly concentrated compounds in a single analysis.
Allows standard-free quantitation of unknowns
The uniform response allows reliable single calibrant quantitation of unknowns
Fundamentally more sensitive than other near-universal detectors>
  • Linear response up to 2 orders of magnitude
  • Dynamic range up to 4 orders of magnitude
  • 10-fold or better LODs than the ELSD
  • LOQs as low as 1-20 ng on a column
Compatible with gradient elution
  • The CAD is compatible with gradient elution when you apply an inverse gradient to maintain the uniform response, unlike the RID
Thermo Scientific™ Vanquish™ Charged Aerosol Detector P Series
Gain confidence, save time, and master compliance
  • Acquire data from multiple detectors without replumbing, reduce contamination by directing unwanted matrix to waste, and improve the quantitative accuracy of early-eluting analytes with the integrated diverter valve
  • Seamlessly transfer your methods from earlier models with built-in method translation tools for consistent cross-instrument performance
  • Full qualification of method parameters for confidence in correct instrument operation
  • Audited control of your instrument flow path supports compliant application flexibility
  • Collect data at four unique power values to quickly identify the ideal linear response
Product offerings
Vanquish CAD P Series
  • Vanquish CAD HP
  • Vanquish CAD FP
Integration with other HPLC/UHPLC instruments
  • Digital and Analog Output options available
Vanquish CAD
  • Vanquish CAD H
  • Vanquish CAD F
Specifications
Communications 1 USB port (USB 2.0, “B” type) 1 USB hub with 3 ports (USB 2.0, “A” type) to connect further Vanquish modules
Data Collection Rate 200 Hz Max.
Detection Mode Records a single power value
Type Standard flow management
For Use With (Equipment) Vanquish HPLC/UHPLC systems
Gas Requirements Nitrogen (recommended) or Compressed Air
GLP Features Predictive Performance functions: Gas filter change interval, service monitoring period, system parameters logged in the Chromeleon Chromatography Data System Audit Trail.
Range Dynamic Range: Up to 4 orders of magnitude
Safety Features Power-up diagnostics, inlet gas over-pressure, high evaporation temp, pump flow shutoff, fluid leak detection and safe leak handling
Temperature Control Settable
Temperature Range Ambient +5° to +100°C
Model Vanquish CAD H