Learn More
Thermo Scientific™ Vanquish™ Dual Pumps
Product Code. 17555456 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Vanquish Dual Pump F
Vanquish™ Dual Pump C
Biocompatible Version:
No
Yes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Biocompatible Version Unit Size
17555456 Vanquish™ Dual Pump C No Each
17861266 Vanquish Dual Pump F Yes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17555456 Supplier Thermo Scientific™ Supplier No. VCP32A01
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Experience greater application flexibility with Thermo Scientific™ Vanquish™ Dual Pumps for HPLC systems.
Obtain the maximum possible application flexibility for Dual LC, Tandem LC or LC-MS, and inverse gradient workflows using Thermo Scientific™ Vanquish™ Dual Pumps. These pumps provide two independent ternary-solvent blending flow streams in one housing. Built-in Thermo Scientific™ SmartFlow™ pumping technology automatically compensates for changing eluent compressibility to ensure excellent flow and gradient precision, independent of eluent composition. This technology gives you unrivaled retention time performance and the highest data confidence to give you freedom in method development and application switching.
View HPLC/UHPLC resources
Dual solvent delivery—highest productivity, fastest return on investment
Vanquish Dual Pumps combine two independent ternary-solvent blending flow streams in a single housing, saving bench space for maximum flexibility including Dual LC, Tandem LC or LC-MS, and inverse gradient workflows.
  • Rely on excellent flow accuracy and precision provided by ultra-precise piston drives
  • Profit from built-in solvent degassing for enhanced flow stability and best detector sensitivity
  • Benefit from outstanding durability for highest system up-time and minimized total cost of ownership
Specifications
Description Vanquish™ Dual Pump C
Biocompatible Version No
Flow Accuracy ±0.1%
Flow Precision <0.05% RSD or <0.01 min SD, whichever is greater
Gradient Formation Dual Low-pressure Gradient Proportioning
Max. Flow Rate 10 mL/min.
Pressure Range 2 to 70 MPa
GLP Features Predictive performance functions for scheduling maintenance procedures based on the actual operating and usage conditions of the pump.
Min. Flow Rate 0.001 mL/min.
Dimensions (HWD) 192 mm × 420 mm × 620 mm (7.6 in. × 16.5 in. × 24.4 in.)
Weight (English) 44 lb.
Weight (Metric) 20 kg
Product Type Serial Dual-Piston Pump