Learn More
Thermo Scientific™ Vanquish™ Fluorescence Detectors
Product Code. 17843025 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Fluorescence Detector C
Wavelength Range:
Excitation: 200-630 nm, Emission: 220-650 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Wavelength Range Unit Size
17843025 Fluorescence Detector C Excitation: 200-630 nm, Emission: 220-650 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17843025 Supplier Thermo Scientific™ Supplier No. VCD50A01
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Experience superior trace-level detection performance with Thermo Scientific™ Vanquish™ Fluorescence Detectors for HPLC.
Get trace detection performance through highly effective stray light suppression with Thermo Scientific™ Vanquish™ Fluorescence Detectors. These detectors reduce thermal effects using temperature-controlled flow cells for increased detection precision. Monitor up to four wavelength pairs simultaneously, or scan your chromatogram for the best emission/excitation wavelengths. Improve detection sensitivity and selectivity by leveraging ultrafast wavelength switching between peaks.
View Fluorescence Detector Resources
  • Acquire data at up to 200 Hz for best support of even the fastest UHPLC separations
  • Optimize your sensitivity using xenon flash lamp frequency and variable emission filter settings
  • Simplify method development or improve existing methods using single spectrum scans or fluorescence field acquisition in excitation, emission, or synchronous mode
  • Improve your cost of ownership with increased lamp lifetime offered by our long-life xenon flash lamp and various lamp operation modes
Specifications
Description Fluorescence Detector C
Data Collection Rate 100 Hz Max.
Detection Mode Single Channel
Detector Type High-Sensitivity Fluorescence
Type Fluorescence Detector
Optical System Two monochromators with concave holographic gratings and elliptic mirrors for highest efficiency in light transmission
PC Connection USB 2.0; 3-port hub to connect further Vanquish modules
Wavelength Range Excitation: 200-630 nm, Emission: 220-650 nm
Dimension 159 mm × 420 mm × 620 mm (6.3 in × 16.5 in × 24.4 in)
Weight (Metric) 21 kg
Voltage 100-240 V AC, 50/60 Hz, max. 150 W/255 VA
Quantity Each