Learn More
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops
Product Code. 18851115 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Vanquish Split Sampler C/CT
Sample Capacity:
250 μL
Biocompatible Version:
No
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Sample Capacity Biocompatible Version Unit Size
18851115 Vanquish Split Sampler C/CT 250 μL No Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18851115 Supplier Thermo Scientific™ Supplier No. 6851.1970
  • ONLINE EXCLUSIVE PRICE
    807.00€
    728.64€ / 1 pièces
    Économisez 78.36€
    9% de réduction
 Restricted Item
Expédition estimée: 21-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops come in a variety of sizes to fit your application requirements..
Automate sample injection into your Thermo Scientific™ Vanquish™ HPLC or UHPLC system with Thermo Scientific™ Vanquish™ Split Sampler Sample Loops. These sample loops are available in 10, 25, 100, 250, 1000 μL sizes to meet your application requirements. Thermo Scientific™ Viper™ Fittings are included with our sample loops to make them easy to install with leak-free connections.
Vanquish Split Sampler Sample Loops use Viper Fittings, which have a ferrule-free tightening design, for virtually zero-dead-volume connections.
TRUSTED_SUSTAINABILITY
Specifications
Type Sample Loop
Sample Capacity 250 μL
Biocompatible Version No
For Use With (Equipment) Vanquish Split Sampler C/CT
Material Stainless Steel
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.