Learn More
Cole-Parmer™ VapLock™ Solvent Collection Systems with 70 mm Rieke 6TPI Cap Size and 20-L Container
Product Code. 16692033 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
PTFE Manifold
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
16692033 PTFE Manifold Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16692033 Supplier Cole-Parmer™ Supplier No. 1202068
  • ONLINE EXCLUSIVE PRICE
    617.00€
    577.22€ / 1 pièces
    Économisez 39.78€
    6% de réduction
Expédition estimée: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
VapLock™ Solvent Collection Systems with 70 mm Rieke 6TPI cap size and 20-L Container are a complete solution to safely collect solvent waste from laboratory instruments
Collect solvent waste from up to 9 effluent streams Minimize exposure to harmful chemical vapors
Warranty
None
Specifications
Description 70 mm Rieke 6 TPI, PTFE manifold, 20 L PE drum
For Use With (Equipment) Chromatography Systems
Type Complete Solvent Collection System
Solvent Channels 9
Material PTFE Manifold