Learn More
WTW™ Variable Wave-length Thermoreactor with 4 Temperature Settings
Product Code. 10159081
Change view
Click to view available options
Holds:
12 Reaction Cuvettes
2 x 12 Reaction Cuvettes
2 x 12 Reaction Cuvettes in 2 heating blocks
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10159081 12 Reaction Cuvettes Each
10041862 2 x 12 Reaction Cuvettes Each
10132342 2 x 12 Reaction Cuvettes in 2 heating blocks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 10159081 Supplier WTW™ Supplier No. 1P211
  • ONLINE EXCLUSIVE PRICE
    1355.00€
    1200.00€ / 1 pièces
    Économisez 155.00€
    11% de réduction
View all promotions
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
For the disintegration of COD, total nitrogen and total phosphorus
Thermoreactors for the disintegration of COD, total nitrogen and total phosphorus, including brief and self-programmed high temperature disintegration up to 170°C. The high reaction temperature over a defined period of time ensures a complete degradation of the sample. The required temperatures and degradation times for the standard parameters are stored in every WTW thermoreactor. In addition, there are different options for self programming and cuvette numbers available.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Thermoreactor
Voltage 230/115 V
Wattage 280 W
Depth (Metric) 315 mm
Electrical Requirements 115/230 V ±15%, 50/60 Hz, 280 W
For Use With (Application) Routine measurements, wastewater
Height (Metric) 185 mm
Holds 12 Reaction Cuvettes
Width (Metric) 256 mm
Frequency 50/60 Hz