Learn More
Veolia Water Systems Veolia Water Systems™ Sterilization Tablet
Product Code. 11964897
Change view
Click to view available options
Quantity:
12/Pk.
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11964897 12/Pk. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11964897 Supplier Veolia Water Systems Supplier No. CT1
 Restricted Item
Expédition estimée: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Reliable and effective chlorine tablets that reduce bio film and protect the system against bacterial growth.
  • CT1 Tablets (pack of 12) - Chlorine tablets
  • Sanitizes tubing or pipe work to remove bacteria
  • Reduces bio film and protect the system against bacterial growth
  • Sterilization tablet
Specifications
Quantity 12/Pk.
compliance-icons compliance-icons
Product Identifier
  • ELGALITE CT-1
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P270-Do not eat, drink or smoke when using this product.
Supplemental information
  • MIXTURE LIST-Contain: troclosene sodium, dihydrate, boric acid