Learn More
Veolia Water Systems™ UV Lamp
Product Code. 12013283
Change view
Click to view available options
For Use With (Equipment):
Bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species
Centra R 60/120, Maxima MK1/MK2, Purelab Option-Q, Purelab Ultra MK1, Purelab Ultra MK2
PURELAB Option-R
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12013283 Bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species Each
11954897 PURELAB Option-R Each
11974897 Centra R 60/120, Maxima MK1/MK2, Purelab Option-Q, Purelab Ultra MK1, Purelab Ultra MK2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12013283 Supplier Veolia Water Systems™ Supplier No. LC21002
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Rich, robust UV lamp that features UV light acting as a bactericide to break down and photo-oxidize organic contaminants to polar or ionized species.
  • UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionized species
  • The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Specifications
For Use With (Application) Used as a bactericide and to break down and photo-oxidize organic contaminats to polar or ionized species.
For Use With (Equipment) Bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species