Learn More
Phenomenex™ Verex™ Preassembled Screw Caps, 9 mm, with Bonded-In Septa, Pre-Slit
Product Code. 31744526 Shop All Phenomenex Products
Change view
Click to view available options
Color:
Black
Blue
Silver
Quantity:
100 Pk.
1000 Pk.
Unit Size:
Pack of 100
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Unit Size
31744526 Blue 100 Pk. Pack of 100
31744037 Blue 1000 Pk. Pack of 1000
31747972 Silver 100 Pk. Pack of 100
31746749 Silver 1000 Pk. Pack of 1000
31744890 Black 100 Pk. Pack of 100
31746529 Black 1000 Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31744526 Supplier Phenomenex™ Supplier No. AR0897212B
Expédition estimée: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Accessories for use with chromatography vials and in various chromatography applications.
  • Analysis: Cleanliness and performance tested as standard
  • Certification: 100% defect-free
  • Configurations: Specialty vials, crimp-top vials, screw-top vials, snap-top vials, VOA/ASE assembled vial kits, VOA/ASE assembled storage kits
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Diameter (Metric) Outer 9 mm
Pre-slit Yes
Sterility Sterile
Septum PTFE/Silicone
Quantity 100 Pk.
Closure Size 9 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap