Learn More
3M™ Versaflo™ Breathing Tube Radiant Heat Cover, BT-927
Product Code. 17267640 Shop All 3M Products
Change view
Click to view available options
Height (Metric):
0.086 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
17267640 0.086 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17267640 Supplier 3M™ Supplier No. 7100099908
  • ONLINE EXCLUSIVE PRICE
    202.00€
    135.76€ / 1 pièces
    Économisez 66.24€
    32% de réduction
Expédition estimée: 15-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Radiant heat resistant, highly durable breathing tube cover for 3M™ Versaflo™ BT-Series Breathing Tubes.
  • Radiant heat resistant, high durability breathing tube cover
  • Lengthens the life of the breathing tube
  • Designed for use with 3M™ Versaflo™ BT -Series Breathing Tubes
TRUSTED_SUSTAINABILITY
Specifications
Color Grey
Length (Metric) 0.355 m
Width (Metric) 0.228 m
Height (Metric) 0.086 m