Learn More
3M™ Versaflo™ Over-visor, M-967
Product Code. 17277640 Shop All 3M Products
Change view
Click to view available options
Headgear Cover
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17277640 Headgear Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17277640 Supplier 3M™ Supplier No. 7100099909
  • ONLINE EXCLUSIVE PRICE
    242.00€
    157.15€ / 1 pièces
    Économisez 84.85€
    35% de réduction
Expédition estimée: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Over-visor to protect against UV/IR, metal sparks and splashes.
  • Gold IR/UV over-visor
  • Protects against UV/IR, and metal sparks and splashes
  • 500 x 250 mm
  • For use with 3M™ Versaflo™ Over-visor Frame, M-961
TRUSTED_SUSTAINABILITY
Specifications
Color Gold
Product Type Headgear Cover
For Use With (Equipment) 3 M™ Versaflo™ Over Visor Frame M 961 N
Quantity 1/Cs.
Length (Metric) 0.503 m
Width (Metric) 0.252 m
Height (Metric) 0.012 m