Learn More
3M™ Versaflo™ Peel-Off Visor Cover
Product Code. 15778997 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15778997 Small Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15778997 Supplier 3M™ Supplier No. 7000027993
  • ONLINE EXCLUSIVE PRICE
    49.00€
    32.91€ / 10 pièces
    Économisez 16.09€
    32% de réduction
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
3M™ Versaflo™ Peel-Off Visor Cover is a self-adhesive cover that protects the visor from contaminants and scratches.
  • For use with 3M™ Versaflo™ S-600, S-700 and S-800 head covers and hoods
  • Self-adhesive visor covers can be easy applied and torn off
  • Protects the visor from contaminants and scratches
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
Product Type Visor Cover
Size Small
For Use With (Equipment) 3M™ Versaflo™ S-600, S-700, and S-800 head covers and hoods