Learn More
3M™ Versaflo™ Powered Air Respirator System Easy Clean Ready Kit, TR-600E ECK
Product Code. 17906870 Shop All 3M Products
Change view
Click to view available options
Color:
Black, White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17906870 Black, White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17906870 Supplier 3M™ Supplier No. 7100258808
  • ONLINE EXCLUSIVE PRICE
    2635.00€
    1865.80€ / 1 pièces
    Économisez 769.20€
    29% de réduction
 Restricted Item
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
3M™ Versaflo™ TR-600 Powered Air Respirators are versatile and easy-to-use respiratory protection against certain gases and vapours and as well as particulates.
  • Respiratory protection against dusts, mists, fumes and other particulate hazards, and a variety of gas and vapour hazards, including organic vapou, acid gas, inorganic gases, ammonia and mercury vapours
  • Three user selectable airflows with loose-fitting and tight-fitting options, enable the user to control flow settings to allow the user to find the setting they find most comfortable
  • Powered Air Respirators automatically adjust airflow to compensate for filter loading to deliver consistent filtered air during use
  • Audible, visual, and vibratory alerts for low airflow and low battery charge-level to keep you prepared and informed
  • High capacity battery option available for longer run time and reduced downtime
  • Available in convienient, easy to buy assemblies and kits
TRUSTED_SUSTAINABILITY
Specifications
Color Black, White
Product Type PAPR Healthcare Systems and Kits
Size Standard
For Use With (Equipment) TR-6712E Filter
Mounting Type Belt-Mounted