Learn More
Brady™ Vinyl Glow-in-the-dark Anti-skid Left Footprint
Product Code. 11721965 Shop All Brady Products
Change view
Click to view available options
Color:
Photoluminescent
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11721965 Photoluminescent Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11721965 Supplier Brady™ Supplier No. 224074
  • ONLINE EXCLUSIVE PRICE
    13.40€
    11.32€ / 1 pièces
    Économisez 2.08€
    15% de réduction
Expédition estimée: 08-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Use the Vinyl Glow-in-the-dark Anti-skid Left Footprint together with the right foot to create a visual pathway to aid a fast exit during power outages and emergency situations. Anti-slip effect.
  • Anti-slip effect.
Specifications
Alternate Part Number GLOW IN THE DARK ANTI SKID FOOT. LEFT
Description Glow-in-the-dark anti-skid footprint - Left foot
For Use With (Application) Use Together With the Right Foot to Create a Visual Pathway
Height (English) 8.27 in.
Height (Metric) 210 mm
Packaging Quantity 1 Ea.
Temperature Range (English) -40°F to +230°F
Temperature Range (Metric) -40°C to +110°C
Thickness 0.6 mm
Width (English) 3.35 in.
Width (Metric) 85 mm
Color Photoluminescent
Material Aluminum