Learn More
Thermo Scientific™ Virtuoso™ Multiple Vial Holder
Product Code. 15291918
Change view
Click to view available options
For Use With (Equipment):
Virtuoso Vial Identification System
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15291918 Virtuoso Vial Identification System Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15291918 Supplier Thermo Scientific™ Supplier No. 60180VT501
Quantity Price
1 16.65€ / 5 pièces
10 14.77€ / 5 pièces
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Print sample data directly on up to 10 vials sequentially for higher throughput using the Thermo Scientific™ Virtuoso™ Multiple Vial Holder.
Print directly on up to 10 vials sequentially for higher throughput using the Thermo Scientific™ Virtuoso™ Multiple Vial Holder for your Virtuoso Vial Identification System. With advanced printing and labeling capabilities, the Virtuoso system ensures accurate vial identification and the integrity of sample data.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Virtuoso
Description Virtuoso Multiple Vial Holder
Device Holds 10 Vials
For Use With (Application) Vial ID and Labelling
For Use With (Equipment) Virtuoso Vial Identification System
No. per Pack 5/Pk.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.