Learn More
Labnet™ Vortemp™ 1550 Shaking Incubator
Product Code. 11422137
Change view
Click to view available options
Holds:
18 x 15 mL or 8 x 50 mL Tubes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11422137 18 x 15 mL or 8 x 50 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 11422137 Supplier Labnet™ Supplier No. S2050A230V
  • ONLINE EXCLUSIVE PRICE
    3522.49€
    3054.76€ / 1 pièces
    Économisez 467.73€
    13% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Vortemp™ 1550 Shaking Incubator - Complete with racks for 50 mL and 15 mL tubes, 230V.
  • Interchangeable and Adjustable Platforms
    • Designed for simultaneous heating and mixing of small samples, the VorTemp™ 1550 shaking incubators are supplied with interchangeable platforms for standard 15 mL and 50 mL conical tubes.
    • Maximum capacity is 8 x 50 mL or 18 x 15 mL.
    • Temperature in the VorTemp™ 1550 is adjustable over a broad range, making the unit useful for a variety of applications, including bacterial cultures and digestions.
    • Mechanical convection provides a uniform and stable environment within the chamber.
    • The VorTemp™ 1550 may also be used in the cold room for sub-ambient applications.
    • Shake and incubate samples simultaneously
    • Intelligent, digital control
  • Microprocessor Controlled Parameters
    • Shaking speed as well as acceleration rates are adjustable.
    • Acceleration can be set to three different levels to protect delicate samples.
    • A microprocessor controls all parameters including temperature and timed operation.
    • Self diagnostic software continuously monitors the unit for proper temperature and shaking speed, alerting the user to any errors.
    • Heating and shaking are controlled by feedback loops which precisely maintain chosen parameters, independent of load or voltage fluctuations.
    • The sturdy construction of the VorTemp™ 1550 gives it stability during high speed shaking.
    • The chamber is insulated to maintain temperature and reduce operating noise.
Specifications
Orbit 3 mm
Speed Range 200 to 950 rpm
Voltage 230 V
Temperature Range (Metric) 5°C to 99°C
Holds 18 x 15 mL or 8 x 50 mL Tubes
Temperature Accuracy ±0.5°C
Dimensions (L x W x H) 26 x 31 x 31 cm (10.2 x 12.2 x 12.2 in.)
Timer Range 1 to 99 min. or Continuous