Learn More
Waring™ Speed Blender
Spécification
Spécification
| Amperage | 1.8 A |
| Voltage | 230 V |
| Electrical Requirements | 230 V, 50/60 Hz, 1.8A |
| Capacity (Metric) | 1 mL |
| Packaging Quantity | 1 Ea. |
| Product Type | Speed Blender |
| Frequency | 50/60 Hz |