Learn More
Waring™ Stainless Steel Mini Container
Product Code. 12839395
Change view
Click to view available options
Volume Range (Metric):
120 to 37 mL
37 to 110 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Range (Metric) Unit Size
12839395 37 to 110 mL Each
12829395 120 to 37 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12839395 Supplier Waring™ Supplier No. 21/MC2
  • ONLINE EXCLUSIVE PRICE
    679.79€
    557.80€ / 1 pièces
    Économisez 121.99€
    17% de réduction
Expédition estimée: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Waring blenders are designed for frequent use
Multi-speed blenders, with a choice of either glass or stainless steel containers, and the unique clover shape for perfect blending action. Waring blenders are designed for frequent use.
Specifications
Volume Range (Metric) 37 to 110 mL
Material Stainless Steel
Includes Stainless Steel Lid
Packaging Quantity 1 Ea.
Product Type Mini Container