Learn More
Bryson Packaging™ Refuse Sacks
Product Code. 30057394
Change view
Click to view available options
Dimensions: 450L x 800W x 975mmH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30057394 Dimensions: 450L x 800W x 975mmH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30057394 Supplier Bryson Packaging™ Supplier No. WDS650525X
  • ONLINE EXCLUSIVE PRICE
    1895.25€
    1703.92€ / 1000 pièces
    Économisez 191.33€
    10% de réduction
Expédition estimée: 12-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
An excellent option for refuse and trash removal from the laboratory. Bryson Packaging™ Refuse Sacks are manufactured from polyethylene.
  • Durable to prevent leaks
  • Provided in a packs of 100
Specifications
Product Type Waste Sack