Learn More
Cytiva Whatman AE98 Cellulose Nitrate Membranes
Product Code. 15843704
Change view
Click to view available options
Dimensions (L x W):
25mm x 50mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15843704 25mm x 50mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15843704 Supplier Cytiva Supplier No. 10549916
  • ONLINE EXCLUSIVE PRICE
    325.00€
    296.39€ / 1 pièces
    Économisez 28.61€
    8% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Recommended for the majority of routine applications
  • Manufactured under strict cleanroom conditions
  • Can directly replace general puspose membrane filters
  • High protein binding
TRUSTED_SUSTAINABILITY
Specifications
Description AE98 Cellulose nitrate membranes
Material Cellulose Nitrate
For Use With (Application) Routine filtration applications
Dimensions (L x W) 25mm x 50mm
compliance-icons
Product Identifier
  • AE98 membrane, cellulose nitrate, 25mm x 50m
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose