Learn More
DWK LIFE SCIENCES™ WHEATON™ Extractors For Soxhlet Extraction Apparatus
Product Code. 12654823 Shop All Wheaton Science Products Products
Change view
Click to view available options
Standard Taper Joint Top:
45/50
55/50
Diameter (English) Inside:
1.57 in.
1.96 in.
Length (English):
8.07 in.
9.84 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Top Diameter (English) Inside Length (English) Unit Size
12654823 45/50 1.57 in. 8.07 in. Each
12481561 55/50 1.96 in. 9.84 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12654823 Supplier DWK LIFE SCIENCES™ Supplier No. 415005
  • ONLINE EXCLUSIVE PRICE
    394.00€
    346.03€ / 1 pièces
    Économisez 47.97€
    12% de réduction
Expédition estimée: 12-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
WHEATON™ Extractors For Soxhlet Extraction Apparatus enable continuous extraction of analytes from a solid into an organic solvent. Wetted parts are made from WHEATON™ 33 low extractable borosilicate glass conforming to ASTM E 438 Type I, Class A requirements.
  • Designed for continuous extraction of analytes from a solid into an organic solvent
  • Wetted parts manufactured from WHEATON 33 low extractable borosilicate glass conforming to ASTM E 438 Type I, Class A requirements
  • Standard taper outer joints provide secure connections to condenser and flask components
  • Compatible with paper, alundum, or fritted glass extraction thimbles
Specifications
For Use With (Equipment) Soxhlet Extraction Apparatus
Product Type Soxhlet Extractor
Capacity (Metric) 250 mL
Material Borosilicate Glass
For Use With (Application) Used for continuous extraction of analytes from a solid into an organic solvent
Certifications/Compliance ASTM E 438
Standard Taper Joint Bottom 24/40
Standard Taper Joint Top 45/50
Diameter (English) Inside 1.57 in.
Diameter (Metric) Inside 4 cm
Length (English) 8.07 in.
Length (Metric) 20.5 cm