Learn More
DWK Life Sciences WHEATON™ Arsine Generators
Product Code. 12435482 Shop All Wheaton Science Products Products
Change view
Click to view available options
Material:
Borosilicate Glass
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12435482 Borosilicate Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12435482 Supplier DWK Life Sciences Supplier No. 180030
  • ONLINE EXCLUSIVE PRICE
    520.00€
    389.46€ / 1 pièces
    Économisez 130.54€
    25% de réduction
Expédition estimée: 22-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
WHEATON™ Arsine Generators are assembled, freestanding apparatus designed for arsenic analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method. Manufactured from WHEATON™ 33 low-extractable borosilicate glass for reliable, reproducible results.
  • Designed for arsenic analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
  • Manufactured from WHEATON 33 low-extractable borosilicate glass
  • Assembled apparatus is freestanding and easy to use
  • Design of catalog No. 02912555 eliminates the ball and socket joint connecting the scrubber and absorber components
Compliance
Conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Product Type Arsine Generator
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Application) For arsine analysis using SDDC method