Learn More
Manutan Wide neck keg, 55 litre capacity
Product Code. 12853669
Encompass_Preferred
Change view
Click to view available options
Capacity (Metric):
55 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12853669 55 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12853669 Supplier Manutan Supplier No. A014708
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High density, 100% food grade, polyethylene kegs with excellent chemical resistance. Integrated rubber ring guarantees air and watertight closing, protecting contents from contaminants and preventing leakage. Rubber sealing ring guarentees air and water tigh seal. Tamper evident lid. Wide neck opening allows easy cleaning and access to contents. UN approved for solids and pastes. Maximum filling temperature 80°C. Cool to 30°C before closing. 42, 55 and 68ltr sizes have handles.
  • Capacity: 55L
  • Dimensions (H x D) : 518mm x 410mm
  • UN specification : X/66kg
Specifications
Capacity (Metric) 55 L