Learn More
Wilmad-LabGlass™ SP Scienceware™ Rotor and Caps for Bruker MAS Probe
Shop All SP Scienceware Products
Click to view available options
Model:
520-1A
528-PP-4VAR
For Use With (Equipment):
For Bruker MAS 3.2mm Probe
For Bruker MAS 7mm Probe
Unit Size:
Each
Description
- Manufactured from high-purity Zirconia
- Caps fitted with O-rings for improved sealing
- MAS Probe and caps compatible for most solid state NMR spectrometers
- Thoroughly inspected before and after precision-machining process ensures no material irregularities
- Spin testing performed to highest specified spinning speed, assuring performance without overspinning the rotor
Specifications
Specifications
| For Use With (Equipment) | For Bruker MAS 7mm Probe |
| Material | Various |
| Description | 5 Rotors, 10 Kel-F Caps and 5 Torlon Caps |
| For Use With (Application) | Solid state NMR spectrometers |
| Model | 528-PP-4VAR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction